siRNA Details
| Gene Symbol: Zap70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289612.1 | NP_001276541.1 | 6 | 1026 | ||
| NM_001289765.1 | NP_001276694.1 | 13 | 1973 | |||
| NM_001289766.1 | NP_001276695.1 | 13 | 1938 | |||
| NM_009539.3 | NP_033565.2 | 13 | 1930 | |||
| GenBank mRNA | AB083210.1 | - | 6 | 1029 | ||
| AB084383.1 | - | 6 | 868 | |||
| AK039883.1 | - | 13 | 1867 | |||
| AK089056.1 | - | 13 | 1902 | |||
| BC029727.1 | - | 13 | 1888 | 4925739 | ||
| U04379.1 | - | 13 | 1884 | |||
| U77667.1 | - | 12 | 1776 | |||

Zap70
Genomic Map