siRNA Details
| Gene Symbol: Zfp13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011747.2 | NP_035877.1 | 4, 5 | 765 | ||
| GenBank mRNA | AK030240.1 | - | 4, 5 | 766 | ||
| BC052082.1 | - | 4, 5 | 689 | 6404424 | ||
| FJ593171.1 | - | 4, 5 | 722 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp13| Gene Symbol: Zfp13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011747.2 | NP_035877.1 | 4, 5 | 765 | ||
| GenBank mRNA | AK030240.1 | - | 4, 5 | 766 | ||
| BC052082.1 | - | 4, 5 | 689 | 6404424 | ||
| FJ593171.1 | - | 4, 5 | 722 | |||