siRNA Details
Gene Symbol: Zfp239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001792.1 | NP_001001792.1 | 4 | 1999 | ||
NM_008616.2 | NP_032642.2 | 3 | 1819 | |||
GenBank mRNA | AK078513.1 | - | 3 | 1813 | ||
AK133328.1 | - | 1658 | ||||
BC003315.1 | - | 1 | 753 | 3155528 | ||
BC080674.1 | - | 1 | 1636 | |||
M32057.1 | - | 1 | 1526 |
