siRNA Details
| Gene Symbol: Zfp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011753.3 | NP_035883.2 | 10 | 1176 | ||
| GenBank mRNA | AK029298.1 | - | 8 | 1441 | ||
| AK082002.1 | - | 10 | 1010 | |||
| AK141740.1 | - | 9 | 1539 | |||
| BC051056.1 | - | 3 | 253 | |||

Zfp26
Genomic Map