siRNA Details
| Gene Symbol: Zfp27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037707.1 | NP_001032796.1 | 5 | 712 | ||
| NM_001285797.1 | NP_001272726.1 | 5 | 984 | |||
| NM_001285798.1 | NP_001272727.1 | 5 | 938 | |||
| NM_011754.2 | NP_035884.2 | 5 | 707 | |||
| GenBank mRNA | AK028500.1 | - | 5 | 939 | ||
| AK135293.1 | - | 5 | 988 | |||
| AK147732.1 | - | 5 | 702 | |||
| BC059037.1 | - | 3 | 651 | 6812360 | ||

Zfp27
Genomic Map