siRNA Details
| Gene Symbol: Zfp35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011755.2 | NP_035885.2 | 3 | 1958 | ||
| GenBank mRNA | AK132703.1 | - | 3 | 1961 | ||
| AK159080.1 | - | 3 | 1912 | |||
| BC138830.1 | - | 3 | 1877 | 8861852 | ||
| BC138832.1 | - | 2 | 1875 | 8861854 | ||
| X17617.1 | - | 3 | 1909 | |||

Zfp35
Genomic Map