siRNA Details
| Gene Symbol: Zfp37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290350.1 | NP_001277279.1 | 4 | 1581 | ||
| NM_001290351.1 | NP_001277280.1 | 4 | 1572 | |||
| NM_001290353.1 | NP_001277282.1 | 4 | 1479 | |||
| NM_009554.4 | NP_033580.3 | 4 | 1569 | |||
| GenBank mRNA | AK032469.1 | - | 4 | 1451 | ||
| BC024863.1 | - | 2 | 1192 | |||
| BC063757.1 | - | 4 | 1453 | 6331110 | ||
| X52533.1 | - | 1 | 867 | |||
| X64413.1 | - | 4 | 1375 | |||
| X89264.1 | - | 5 | 1481 | |||

Zfp37
Genomic Map