siRNA Details
| Gene Symbol: Zfp40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009555.2 | NP_033581.2 | 6 | 1804 | ||
| GenBank mRNA | AK049894.1 | - | 6 | 1809 | ||
| BC061205.1 | - | 6 | 1773 | 30248495 | ||
| BC065097.1 | - | 6 | 1797 | |||
| D10632.1 | - | 2 | 1486 | |||

Zfp40
Genomic Map