siRNA Details
| Gene Symbol: Zfp42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009556.3 | NP_033582.2 | 4 | 1039 | ||
| GenBank mRNA | AB221555.1 | - | 1 | 638 | ||
| BC053399.1 | - | 4 | 1012 | 30031349 | ||
| BC064785.1 | - | 5 | 1070 | |||
| BC098467.1 | - | 4 | 974 | 30008039 | ||
| M28382.1 | - | 4 | 1000 | |||

Zfp42
Genomic Map