siRNA Details
| Gene Symbol: Zfp51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133677.1 | - | 5 | 496 | ||
| AK154157.1 | - | 5 | 502 | |||
| CK390565.1 | - | 5 | 485 | |||
| X74855.1 | - | 5 | 527 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp51| Gene Symbol: Zfp51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133677.1 | - | 5 | 496 | ||
| AK154157.1 | - | 5 | 502 | |||
| CK390565.1 | - | 5 | 485 | |||
| X74855.1 | - | 5 | 527 | |||