siRNA Details
| Gene Symbol: Zfp61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009561.2 | NP_033587.2 | 7 | 1650 | ||
| GenBank mRNA | AK148158.1 | - | 7 | 1468 | ||
| AK149960.1 | - | 7 | 1445 | |||
| BC006618.1 | - | 7 | 1689 | 3967527 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp61| Gene Symbol: Zfp61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009561.2 | NP_033587.2 | 7 | 1650 | ||
| GenBank mRNA | AK148158.1 | - | 7 | 1468 | ||
| AK149960.1 | - | 7 | 1445 | |||
| BC006618.1 | - | 7 | 1689 | 3967527 | ||