siRNA Details
| Gene Symbol: Zfp93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009567.4 | NP_033593.1 | 4 | 882 | ||
| GenBank mRNA | AK028905.1 | - | 4 | 720 | ||
| AK040325.1 | - | 4 | 813 | |||
| AK132229.1 | - | 4 | 883 | |||
| AK146095.1 | - | 4 | 873 | |||
| AK165630.1 | - | 4 | 843 | |||
| BC003776.1 | - | 605 | 3496816 | |||
| BC061487.1 | - | 3 | 663 | 5349153 | ||
| U46186.1 | - | 4 | 874 | |||

Zfp93
Genomic Map