siRNA Details
| Gene Symbol: Zfp94 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199321.1 | NP_001186250.1 | 6 | 1262 | ||
| NM_009568.3 | NP_033594.1 | 5 | 1223 | |||
| GenBank mRNA | AK138205.1 | - | 6 | 1203 | ||
| BC005437.1 | - | 5 | 1223 | 3591835 | ||
| BC095945.1 | - | 4 | 1043 | 4936327 | ||
| U46187.1 | - | 5 | 1166 | |||

Zfp94
Genomic Map