siRNA Details
| Gene Symbol: Zkscan5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167944.1 | NP_001161416.1 | 3, 4 | 1889 | ||
| NM_016683.2 | NP_057892.2 | 5, 6 | 2108 | |||
| GenBank mRNA | AB221634.1 | - | 5, 6 | 1372 | ||
| AK173086.1 | - | 5, 6 | 2103 | |||
| BC060248.1 | - | 3, 4 | 1784 | 5715760 | ||
| U62907.1 | - | 5, 6 | 1432 | |||

Zkscan5
Genomic Map