siRNA Details
| Gene Symbol: Zscan12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016684.2 | NP_057893.2 | 4 | 886 | ||
| GenBank mRNA | AK129138.1 | - | 4 | 886 | ||
| AK139837.1 | - | 5 | 1209 | |||
| AK142005.1 | - | 4 | 865 | |||
| BC027041.1 | - | 3 | 844 | 5364568 | ||
| U62908.1 | - | 3 | 1195 | |||

Zscan12
Genomic Map