siRNA Details
| Gene Symbol: Zfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011767.2 | NP_035897.2 | 3, 4 | 485 | ||
| GenBank mRNA | AF071059.1 | - | 3, 4 | 483 | ||
| BC038599.1 | - | 3, 4 | 489 | |||
| BC058570.1 | - | 3, 4 | 485 | |||
| BC094220.1 | - | 2, 3 | 373 | |||
| BC141139.1 | - | 3, 4 | 469 | 9055751 | ||

Zfr
Genomic Map