siRNA Details
| Gene Symbol: Zhx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042438.2 | NP_001035903.1 | 4 | 1415 | ||
| NM_009572.4 | NP_033598.2 | 3 | 1289 | |||
| GenBank mRNA | AK051410.1 | - | 3 | 1278 | ||
| AK053191.1 | - | 3 | 999 | |||
| AK220508.1 | - | 3 | 1250 | |||
| BC004723.1 | - | 4 | 1121 | |||
| BC054543.1 | - | 2 | 873 | 6406286 | ||
| Z54200.1 | - | 3 | 1000 | |||

Zhx1
Genomic Map