siRNA Details
| Gene Symbol: Zic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009573.3 | NP_033599.2 | 1 | 1083 | ||
| GenBank mRNA | AK136977.1 | - | 1 | 308 | ||
| AK141883.1 | - | 1 | 277 | |||
| AK159769.1 | - | 1 | 1055 | |||
| BC050889.1 | - | 1 | 321 | 6418777 | ||
| BC060247.1 | - | 2 | 663 | 6813617 | ||
| BC063247.1 | - | 1 | 1045 | 5694793 | ||
| D32167.1 | - | 1 | 831 | |||

Zic1
Genomic Map