siRNA Details
| Gene Symbol: Zic4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009576.2 | NP_033602.2 | 4 | 1197 | ||
| GenBank mRNA | AK028305.1 | - | 2 | 1000 | ||
| AK035908.1 | - | 2 | 1001 | |||
| AK078127.1 | - | 2 | 314 | |||
| D78174.1 | - | 2 | 1450 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zic4| Gene Symbol: Zic4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009576.2 | NP_033602.2 | 4 | 1197 | ||
| GenBank mRNA | AK028305.1 | - | 2 | 1000 | ||
| AK035908.1 | - | 2 | 1001 | |||
| AK078127.1 | - | 2 | 314 | |||
| D78174.1 | - | 2 | 1450 | |||