siRNA Details
| Gene Symbol: Ikzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036833.1 | - | 5, 6 | 856 | ||
| AK040268.1 | - | 4, 5 | 590 | |||
| AK077815.1 | - | 5, 6 | 1154 | |||
| AK154175.1 | - | 5, 6 | 879 | |||
| AK220235.1 | - | 5, 6 | 823 | |||
| BC138789.1 | - | 4, 5 | 388 | 8861811 | ||
| L03547.1 | - | 4, 5 | 373 | |||
| S74708.1 | - | 2, 3 | 434 | |||

Ikzf1
Genomic Map