siRNA Details
Gene Symbol: Ikzf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011770.4 | NP_035900.2 | 7 | 1075 | ||
GenBank mRNA | AF044257.1 | - | 7 | 970 | ||
AK047925.1 | - | 7 | 937 | |||
BC138606.1 | - | 7 | 915 | 8861626 | ||
BC138608.1 | - | 7 | 915 | 8861628 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106916, XM_006245120, XM_008767217, XM_008767218, XM_006245119, and others.
Panther Classification:
Gene Ontology Categories:
