siRNA Details
Gene Symbol: Ikzf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011771.1 | NP_035901.1 | 8 | 1149 | ||
GenBank mRNA | AF001293.1 | - | 7 | 868 | ||
AK038043.1 | - | 2 | 1612 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ikzf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011771.1 | NP_035901.1 | 8 | 1149 | ||
GenBank mRNA | AF001293.1 | - | 7 | 868 | ||
AK038043.1 | - | 2 | 1612 |