siRNA Details
Gene Symbol: Ikzf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011771.1 | NP_035901.1 | 4 | 544 | ||
GenBank mRNA | AF001293.1 | - | 3 | 263 | ||
AI549961.1 | - | 4 | 290 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ikzf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011771.1 | NP_035901.1 | 4 | 544 | ||
GenBank mRNA | AF001293.1 | - | 3 | 263 | ||
AI549961.1 | - | 4 | 290 |