siRNA Details
| Gene Symbol: Ikzf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011772.2 | NP_035902.2 | 8 | 1866 | ||
| GenBank mRNA | AB017615.1 | - | 7 | 2273 | ||
| AK089522.1 | - | 8 | 1700 | |||
| AK165740.1 | - | 2131 | ||||
| AK170453.1 | - | 9 | 1790 | |||
| BC141286.1 | - | 7 | 1964 | 9055898 | ||
| BC145614.1 | - | 7 | 1950 | |||

Ikzf4
Genomic Map