siRNA Details
| Gene Symbol: Ikzf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011772.2 | NP_035902.2 | 2 | 335 | ||
| GenBank mRNA | AB017615.1 | - | 1 | 742 | ||
| AK089522.1 | - | 2 | 169 | |||
| AK165740.1 | - | 599 | ||||
| AK170453.1 | - | 2 | 187 | |||
| BC030070.1 | - | 1 | 533 | |||
| BC141286.1 | - | 1 | 433 | 9055898 | ||
| BC145614.1 | - | 1 | 433 | |||

Ikzf4
Genomic Map