siRNA Details
| Gene Symbol: Slc30a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011773.3 | NP_035903.2 | 7 | 1075 | ||
| GenBank mRNA | AK144568.1 | - | 7 | 1059 | ||
| AK165796.1 | - | 7 | 1038 | |||
| BC066199.1 | - | 7 | 1057 | 30476359 | ||
| U76007.1 | - | 7 | 1033 | |||

Slc30a3
Genomic Map