siRNA Details
| Gene Symbol: Adam25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011781.2 | NP_035911.2 | 2 | 2096 | ||
| GenBank mRNA | AF167403.1 | - | 2096 | |||
| AK016581.1 | - | 2 | 2094 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam25| Gene Symbol: Adam25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011781.2 | NP_035911.2 | 2 | 2096 | ||
| GenBank mRNA | AF167403.1 | - | 2096 | |||
| AK016581.1 | - | 2 | 2094 | |||