siRNA Details
| Gene Symbol: Adamts5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011782.2 | NP_035912.2 | 3 | 2197 | ||
| GenBank mRNA | AF140673.1 | - | 3 | 1356 | ||
| AK044746.1 | - | 3 | 419 | |||
| AK082475.1 | - | 3 | 419 | |||
| BC138619.1 | - | 3 | 1381 | 8861639 | ||
| BC138620.1 | - | 3 | 1381 | 8861640 | ||

Adamts5
Genomic Map