siRNA Details
| Gene Symbol: JAG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002226.4 | NP_002217.3 | 13 | 2070 | ||
| NM_145159.2 | NP_660142.1 | 12 | 1956 | |||
| GenBank mRNA | AF003521.1 | - | 13 | 1698 | ||
| AF020201.1 | - | 13 | 1717 | |||
| AF029778.1 | - | 13 | 2070 | |||
| AF029779.1 | - | 12 | 1956 | |||
| BC032053.1 | - | 13 | 1734 | |||
| U69180.1 | - | 4 | 328 | |||
| Y14330.1 | - | 13 | 1621 | |||

JAG2
Genomic Map