siRNA Details
| Gene Symbol: Akt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011785.3 | NP_035915.3 | 10 | 1046 | ||
| GenBank mRNA | AF124142.1 | - | 10 | 1036 | ||
| AK052953.1 | - | 2 | 158 | |||
| BC066861.1 | - | 10 | 1046 | 30089997 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akt3| Gene Symbol: Akt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011785.3 | NP_035915.3 | 10 | 1046 | ||
| GenBank mRNA | AF124142.1 | - | 10 | 1036 | ||
| AK052953.1 | - | 2 | 158 | |||
| BC066861.1 | - | 10 | 1046 | 30089997 | ||