siRNA Details
| Gene Symbol: Aloxe3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011786.2 | NP_035916.2 | 14 | 1933 | ||
| GenBank mRNA | BC116258.1 | - | 14 | 1932 | 40093733 | |
| BC116259.1 | - | 14 | 1932 | 40093735 | ||
| Y14695.1 | - | 14 | 1933 | |||

Aloxe3
Genomic Map