siRNA Details
| Gene Symbol: Bves | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024285.2 | NP_077247.1 | 6 | 863 | ||
| GenBank mRNA | AF124510.1 | - | 5 | 724 | ||
| AF204174.1 | - | 6 | 861 | |||
| BC132018.1 | - | 5 | 753 | 40130295 | ||
| BC132044.1 | - | 5 | 753 | 40130321 | ||

Bves
Genomic Map