siRNA Details
| Gene Symbol: Def6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027185.3 | NP_081461.2 | 3 | 406 | ||
| GenBank mRNA | AF329497.1 | - | 3 | 341 | ||
| AK038050.1 | - | 3 | 377 | |||
| AK088460.1 | - | 3 | 355 | |||
| AY241695.1 | - | 3 | 341 | |||
| BC120500.1 | - | 3 | 406 | 40129423 | ||
| BC131953.1 | - | 3 | 406 | 40130230 | ||
| X96705.1 | - | 3 | 298 | |||

Def6
Genomic Map