siRNA Details
| Gene Symbol: Dlg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011807.3 | NP_035937.2 | 8 | 1003 | ||
| GenBank mRNA | AF388675.1 | - | 8 | 782 | ||
| AK046525.1 | - | 7 | 1043 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dlg2| Gene Symbol: Dlg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011807.3 | NP_035937.2 | 8 | 1003 | ||
| GenBank mRNA | AF388675.1 | - | 8 | 782 | ||
| AK046525.1 | - | 7 | 1043 | |||