siRNA Details
| Gene Symbol: Fbln5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011812.4 | NP_035942.1 | 3, 4 | 490 | ||
| GenBank mRNA | AF112151.1 | - | 3, 4 | 440 | ||
| AK084895.1 | - | 3, 4 | 331 | |||
| AK085170.1 | - | 3, 4 | 320 | |||
| AK090129.1 | - | 3, 4 | 429 | |||
| AK159189.1 | - | 3, 4 | 253 | |||
| AK159471.1 | - | 3, 4 | 473 | |||
| AK159960.1 | - | 3, 4 | 460 | |||
| AK165018.1 | - | 3, 4 | 283 | |||
| BC006636.1 | - | 3, 4 | 180 | 3482574 | ||

Fbln5
Genomic Map