siRNA Details
| Gene Symbol: Gpc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079844.2 | NP_001073313.1 | 3 | 1080 | ||
| NM_011821.3 | NP_035951.1 | 3 | 1080 | |||
| GenBank mRNA | AF105268.1 | - | 4 | 803 | ||
| AK032866.1 | - | 3 | 501 | |||
| AK132531.1 | - | 3 | 817 | |||
| AK135442.1 | - | 3 | 1062 | |||
| AK149312.1 | - | 3 | 847 | |||
| AK164668.1 | - | 3 | 1082 | |||
| BC023448.1 | - | 3 | 494 | 5008374 | ||

Gpc6
Genomic Map