siRNA Details
Gene Symbol: Hs2st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011828.3 | NP_035958.3 | 2 | 630 | ||
GenBank mRNA | AF060178.1 | - | 2 | 308 | ||
AF169243.1 | - | 2 | 297 | |||
AK088698.1 | - | 2 | 606 | |||
AK165112.1 | - | 2 | 596 | |||
BC025443.1 | - | 2 | 558 | 5068428 | ||
BC059008.1 | - | 2 | 387 | 6849136 |
