siRNA Details
| Gene Symbol: Insrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011832.2 | NP_035962.2 | 7 | 1930 | ||
| GenBank mRNA | AB007135.1 | - | 7 | 1819 | ||
| AK081512.1 | - | 7 | 1917 | |||
| AK142578.1 | - | 7 | 1931 | |||
| BC137855.1 | - | 7 | 1787 | 8860874 | ||

Insrr
Genomic Map