siRNA Details
| Gene Symbol: Aadat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011834.2 | NP_035964.1 | 4 | 524 | ||
| GenBank mRNA | AF072376.1 | - | 4 | 524 | ||
| AK075578.1 | - | 4 | 530 | |||
| AK144042.1 | - | 4 | 477 | |||
| BC012637.1 | - | 4 | 527 | 4160508 | ||

Aadat
Genomic Map