siRNA Details
| Gene Symbol: Lamc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011836.3 | NP_035966.2 | 23, 24 | 4022 | ||
| GenBank mRNA | AF079520.1 | - | 23, 24 | 4045 | ||
| AF083372.1 | - | 23, 24 | 3959 | |||
| AK132948.1 | - | 20, 21 | 3009 | |||
| BC005535.1 | - | 6, 7 | 767 | |||
| BC096366.1 | - | 23, 24 | 4022 | 2865309 | ||

Lamc3
Genomic Map