siRNA Details
| Gene Symbol: Ly6h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135688.1 | NP_001129160.1 | 4 | 451 | ||
| NM_001135689.1 | NP_001129161.1 | 4 | 365 | |||
| NM_011837.3 | NP_035967.1 | 4 | 728 | |||
| GenBank mRNA | AF127091.1 | - | 4 | 361 | ||
| AK021002.1 | - | 4 | 405 | |||
| AK034884.1 | - | 368 | ||||
| BB627076.1 | - | 4 | 368 | |||
| BC028758.1 | - | 4 | 362 | 1245940 | ||
| BM943584.1 | - | 4 | 451 | |||
| BM948855.1 | - | 4 | 728 | |||
| CX238172.1 | - | 4 | 431 | |||

Ly6h
Genomic Map