siRNA Details
| Gene Symbol: Nek4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF099067.1 | - | 7 | 1240 | ||
| AJ223071.1 | - | 7 | 1330 | |||
| AK078809.1 | - | 7 | 1352 | |||
| AK132803.1 | - | 7 | 1268 | |||
| AK147381.1 | - | 7 | 1318 | |||
| AK149584.1 | - | 7 | 1268 | |||
| BC057939.1 | - | 7 | 1242 | 5325558 | ||
| CN526970.1 | - | 2 | 236 | |||
| Y09234.1 | - | 7 | 1330 | |||

Nek4
Genomic Map