siRNA Details
| Gene Symbol: Nr2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013708.4 | NP_038736.1 | 8 | 1797 | ||
| GenBank mRNA | AF204053.1 | - | 8 | 1797 | ||
| AK045788.1 | - | 6 | 1540 | |||
| AK045859.1 | - | 6 | 1540 | |||
| BC047150.1 | - | 5 | 2484 | |||

Nr2e3
Genomic Map