siRNA Details
| Gene Symbol: Tenm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011858.3 | NP_035988.2 | 31 | 6612 | ||
| GenBank mRNA | AB025413.1 | - | 29 | 6445 | ||
| AF059485.1 | - | 31 | 7002 | |||
| AK122490.1 | - | 5 | 1539 | |||
| AK147329.1 | - | 31 | 6613 | |||
| AK147579.1 | - | 31 | 6912 | |||

Tenm4
Genomic Map