siRNA Details
| Gene Symbol: Eci2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110331.1 | NP_001103801.1 | 4, 5 | 590 | ||
| NM_011868.3 | NP_035998.2 | 4, 5 | 774 | |||
| GenBank mRNA | AF153613.1 | - | 4, 5 | 746 | ||
| AK009478.1 | - | 4, 5 | 574 | |||
| AK170778.1 | - | 4, 5 | 559 | |||
| AK218002.1 | - | 1, 2 | 49 | |||
| BC001983.1 | - | 4, 5 | 484 | 3591615 | ||
| CB318094.1 | - | 4, 5 | 571 | |||

Eci2
Genomic Map