siRNA Details
| Gene Symbol: Prkra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011871.2 | NP_036001.1 | 2, 3 | 379 | ||
| GenBank mRNA | AK012798.1 | - | 2, 3 | 365 | ||
| AK155112.1 | - | 2, 3 | 403 | |||
| BC011311.1 | - | 2, 3 | 387 | 3985758 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkra| Gene Symbol: Prkra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011871.2 | NP_036001.1 | 2, 3 | 379 | ||
| GenBank mRNA | AK012798.1 | - | 2, 3 | 365 | ||
| AK155112.1 | - | 2, 3 | 403 | |||
| BC011311.1 | - | 2, 3 | 387 | 3985758 | ||