siRNA Details
| Gene Symbol: Klk7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011872.2 | NP_036002.1 | 4 | 360 | ||
| GenBank mRNA | AB008371.1 | - | 4 | 337 | ||
| AF124299.1 | - | 4 | 362 | |||
| AK029477.1 | - | 4 | 360 | |||
| AK077406.1 | - | 4 | 360 | |||
| BC027823.1 | - | 4 | 521 | 5322691 | ||

Klk7
Genomic Map