siRNA Details
| Gene Symbol: Tiam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122998.1 | NP_001116470.1 | 3 | 373 | ||
| NM_011878.2 | NP_036008.2 | 3 | 357 | |||
| GenBank mRNA | AB022915.1 | - | 3 | 358 | ||
| AK129474.1 | - | 3 | 276 | |||
| AK158137.1 | - | 3 | 336 | |||
| BC067048.1 | - | 3 | 373 | 30535152 | ||
| BC079600.1 | - | 3 | 262 | 6853027 | ||

Tiam2
Genomic Map