siRNA Details
| Gene Symbol: Rai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103367.1 | NP_001096837.1 | 2 | 1541 | ||
| NM_198409.3 | NP_940801.2 | 2 | 1619 | |||
| GenBank mRNA | AK143442.1 | - | 2 | 1509 | ||
| BC061184.1 | - | 2 | 1545 | 30300326 | ||
| X76652.1 | - | 1 | 1285 | |||

Rai2
Genomic Map