siRNA Details
Gene Symbol: Grk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011881.3 | NP_036011.3 | 1 | 128 | ||
GenBank mRNA | BC075722.1 | - | 1 | 103 | ||
BC096611.1 | - | 1 | 128 | 5357325 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Grk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011881.3 | NP_036011.3 | 1 | 128 | ||
GenBank mRNA | BC075722.1 | - | 1 | 103 | ||
BC096611.1 | - | 1 | 128 | 5357325 |